Anti-AKT3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25F6.F6.D8]

Supplier: Rockland Immunochemicals
200-326-E75 200-326-E75
CA200-326-E75EA 781.53 CAD
RL200-326-E75 CA200-326-E75
Anti-AKT3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 25F6.F6.D8]
Antibodies
Anti-AKT3 APC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: Flow Cytometry: 1.0 µg/ml;
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 25F6.F6.D8
Conjugation: APC (Allophycocyanin)
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR