Anti-SETDB2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-N99
CA200-401-N99EA 779.7 CAD
CA200-401-N99
Anti-SETDB2 Rabbit Polyclonal Antibody
Antibodies
Anti-Setdb2 antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80.6 kDa in size corresponding to Setdb2 by western blotting.

Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:10000 - 1:100000

Type: Primary
Antigen: SETDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR