Anti-Streptavidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
200-403-095
CA200-403-095EA 935.3 CAD
CA200-403-095
Anti-Streptavidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
This product has been assayed against 1.0 µg of Streptavidin in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:2000-1:10000
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Tag
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR