Anti-TRP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-403-139S 200-403-139S
CA10037-012EA 254.9 CAD
RL200-403-139S CA10037-012
Anti-TRP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Trypsin Antibody is suitable for western blotting and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:14000; Western Blot: 1:1000 - 1:5000;
Do not add Sodium Azide!

Type: Primary
Antigen: TRP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR