Anti-ADA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-403-140S 200-403-140S
CA10037-016EA 254.9 CAD
RL200-403-140S CA10037-016
Anti-ADA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Adenosine deaminase has been assayed against 1.0 µg of Adenosine Deaminase (Calf Spleen) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR