Anti-HAAP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
200-4340S
CA10037-038EA 254.9 CAD
CA10037-038
Anti-HAAP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Acid Phosphatase is suitable for ELISA, western blot.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Do not add Sodium Azide!

Type: Primary
Antigen: HAAP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Potato
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR