Anti-IL2RA Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: PC61]

Supplier: Rockland Immunochemicals
200-508-M73 200-508-M73
CA10037-158EA 833.86 CAD
RL200-508-M73 CA10037-158
Anti-IL2RA Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: PC61]
Antibodies
Anti-CD25 is useful for Immunofluorescence and Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.2-1 µg/1x10^6 cells
Do not freeze! Light sensitive!

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: PC61
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR