Anti-Fcgr3 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]

Supplier: Rockland Immunochemicals
200-508-N80 200-508-N80
CA200-508-N80EA 1033.29 CAD
CA200-508-N80 RL200-508-N80
Anti-Fcgr3 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]
Antibodies
Anti-CD16/32 is useful for Immunoprecipitation and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: Fcgr3
Clonality: Monoclonal
Clone: 93
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR