Anti-IL2RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6D5]

Supplier: Rockland Immunochemicals
200-526-M73 200-526-M73
CA10037-166EA 1075.19 CAD
RL200-526-M73 CA10037-166
Anti-IL2RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6D5]
Antibodies
Anti-CD25 is useful for Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.1-0.5 µg/1x10^6 cells
Do not freeze!

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 6D5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR