Anti-DAO Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-603-143S 200-603-143S
CA10037-170EA 254.9 CAD
CA10037-170 RL200-603-143S
Anti-DAO Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-D-Amino Acid Oxidase has been assayed against 1.0 µg of D-Amino Acid Oxidase (Pig Kidney) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: DAO
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR