Anti-TCRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H57-597]

Supplier: Rockland Immunochemicals
200-B02-N92 200-B02-N92
CA200-B02-N92EA 691.19 CAD
RL200-B02-N92 CA200-B02-N92
Anti-TCRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H57-597]
Antibodies
Anti-TCRbeta is useful for Immunofluorescence, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: TCRB
Clonality: Monoclonal
Clone: H57-597
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR