Anti-TCRB Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: H57-597]

Supplier: Rockland Immunochemical
200-B26-N92
CA200-B26-N92EA 343.93 CAD
CA200-B26-N92
Anti-TCRB Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: H57-597]
Antibodies
Anti-TCRbeta is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: TCRB
Clonality: Monoclonal
Clone: H57-597
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR