Anti-USP7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-482 600-401-482
CA600-401-482EA 971.21 CAD
RL600-401-482 CA600-401-482
Anti-USP7 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 130 kDa in size corresponding to HAUSP protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: USP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR