Anti-JAK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J46 600-401-J46
CA10037-260EA 932.37 CAD
RL600-401-J46 CA10037-260
Anti-JAK2 Rabbit Polyclonal Antibody
Antibodies
Anti-JAK2 antibody is useful for ELISA, immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~130kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1µg/ml

Type: Primary
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR