Anti-BLM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-J60 600-401-J60
CA10037-288EA 932.37 CAD
CA10037-288 RL600-401-J60
Anti-BLM Rabbit Polyclonal Antibody
Antibodies
Anti-BLM pT99 antibody is useful for ELISA, Immunohistochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~159kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1µg/ml

Type: Primary
Antigen: BLM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR