Stop Solution, Enzo Life Sciences
Supplier: Enzo Life Sciences
80-0247
CA95043-924EA 558.14 CAD
CA95043-924
Stop Solution, Enzo Life Sciences
Assays ELISAs
Assay stop solution for AP EIA/ELISA
  • Liquid in form
This is a solution of TRIS-sodium phosphate in water.