12(S)-HETE ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-050
89141-102EA 967.13 CAD
89141-102
12(S)-HETE ELISA Kit, Enzo Life Sciences
Assays ELISAs
Sensitive ELISA kit enabling detection of 12(S)-HETE in culture supernatants and plasma from any species.
For the quantitative determination of 12(S)-HETE in culture supernatants and plasma from any species. Cited sample types include serum, renal interstitial fluid and tissue.
The 12(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Fast, accurate, and reliable results are obtained with ready-to-use liquid color-coded reagents that reduce error.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-300 (5x96 wells).