ImmunoSet® HSP25 (rodent), ELISA Development Set, Enzo Life Sciences
Supplier: Enzo Life Sciences
ImmunoSet®
ADI-960-075
89141-788EA 939.61 CAD
89141-788
ImmunoSet® HSP25 (rodent), ELISA Development Set, Enzo Life Sciences
Assays ELISAs
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of HSP25 in cell lysates of mouse and rat origin.
The ImmunoSet® HSP25 (rodent), ELISA development set contains sufficient reagents for the development of 5 x 96 well ELISA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own ELISA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.
Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.