Human CXCL7 PicoKine ELISA Kit, Boster
Supplier: Boster
PicoKine
EK0729
10207-826EA 1432.87 CAD
10207-826
Human CXCL7 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Human CXCL7
Ordering information: Kits include 96-well plate precoated with anti- human CXCL7 antibody 1; ST0000-10 Lyophilized recombinant human CXCL7 standard 10ng/tubex2; AR1107 Biotinylated anti- human CXCL7 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.