Mouse PECAM-1/CD31 PicoKine ELISA Kit, Boster
Supplier: Boster Biological Technology
EK0874
10207-604EA 1630.88 CAD
10207-604
Mouse PECAM-1/CD31 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Mouse PECAM-1/CD31
Ordering information: Kits include 96-well plate precoated with anti- mouse PECAM-1/CD31 antibody 1; ST0000-10 Lyophilized recombinant mouse PECAM-1/CD31 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse PECAM-1/CD31 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.