Human Angiostatin K1-3 PicoKine ELISA Kit, Boster
Supplier: Boster
PicoKine
EK0905
10205-180EA 1669.71 CAD
10205-180
Human Angiostatin K1-3 PicoKine ELISA Kit, Boster
Assays ELISAs
Sandwich ELISA kit of Quantitative Detection for Human Angiostatin K1-3
Ordering information: Kits include 96-well plate precoated with anti- human Angiostatin K1-3 antibody 1; ST0000-100 Lyophilized recombinant human Angiostatin K1-3 standard 100ng/tubex2; AR1107 Biotinylated anti- human Angiostatin K1-3 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.