Inspection Mirror, Micronova
Catalog #: 10753-250
Supplier: Micronova
Small Business Enterprise
Price
$678.59
Log in for availability
Specifications
- Description:Inspection Mirror, Electropolished Stainless Steel
- Cat. no.:10753-250
Specifications
About this item
A highly polished stainless steel mirror for inspecting hard to reach areas.
- Autoclavable
This 6 x 6" mirror features a swivel connection allowing it to be attached to any SSU series handle. This allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned.
The optional flashlight attachment is ideal for use in tanks and other low light applications.
Customers Who Bought This Also Bought