Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
New Product
Certificates
About this item
Filter dispensers are ideal for high throughput microbiology testing laboratories looking for more ergonomic, faster ways to handle individual membranes. The Sentino® filter dispenser is designed to aseptically dispense one disc of membrane at the touch of a button.
- Easy to load without jamming. Guide plates assist loading and keep refill web in alignment during use
- Soft touch keypad for aseptically dispensing membrane discs
- Designed to hold 200 membrane discs
- Compact, portable design with miniaturised refill pack
- Use plugged into main power source or charge battery for use anywhere. Lights indicate when battery charge is running low
- Easy to clean
The Sentino® filter dispenser can be used for: Drinking and wastewater analysis, environmental and recreational water monitoring, personal care product manufacturing, beverage monitoring, pharmaceutical and industrial water system monitoring, pharmaceutical raw materials, intermediates and final product.
Related Information
Pall Laboratory Microbiology QC Beverage Pall Laboratory Microbiology QC products.pdf
Promotion Material
Pall Laboratory Microbiology QC Products Catalog Pall Laboratory Simplifying Sample Preparation for Research Laboratories
Product Literature
The Importance of Filtration in the Environmental DNA (eDNA) World
Application Notes
Customers Who Bought This Also Bought