Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Infrared Lamps

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/32/18989432.jpg

Conveniently and reliably heat or dry samples from evaporating dishes with an infrared lamp. These heat lamps commonly feature incandescent bulbs which are able to produce the required radiation. Measuring the infrared absorption characteristics of certain liquids and gases through chemical analysis is a conventional use of the devices. Adjustable support arms allow for maneuverability in positioning the infrared lamp bulb, while the convenient on/off switch location and long power cord make it easy to use.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (470005-592)

Supplier:  VWR
Description:   Recommended bulb is clear 150W.

Supplier:  Walter Stern
Description:   Provides a convenient and reliable source for near-infrared rays for heating and drying
Catalog Number: (470005-594)

Supplier:  GENERAL ELECTRIC CO.
Description:   Replacement bulbs for infrared lamp and reflector.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 3  of 3
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping