Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • SBE (4)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

TLC Development Tanks

https://ca.vwr.com/stibo/smallweb/std.lang.all/98/89/18989889.jpg

TLC development tanks are constructed of sturdy glass with beveled edges to eliminate sharp hazards. Clear sides offer an unobstructed view of the developing tank contents. Different tanks are available with lids in a variety of styles, including latch, nonslip, and standard variants. Plate size compatibility varies between different models, with development tanks offered in both rectangular and cylindrical formats with varied internal dimensions. Some rectangular models feature a raised edge along the inside bottom to allow for the simultaneous development of two plates.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"12"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Sturdy molded glass developing tanks feature clear sides to allow unobstructed visual inspection of TLC plates up to 20x20cm (77/8
Supplier:  Wilmad-LabGlass
Description:   These heavy wall cylindrical borosilicate glass chromatography tanks are supplied complete with Standard Taper ground glass stoppers.
Supplier:  Wilmad-LabGlass
Description:   These borosilicate glass cylindrical thin layer chromatography (TLC) developing chambers have flat bottoms and molded plastic caps.
Catalog Number: (KT416180-1020)

Supplier:  DWK Life Sciences (KIMBLE®)
Description:   The standard 20 x 20 cm TLC plate tank is difficult to use with 10 x 10 cm plates, so the “SHORTY” tank was developed
Supplier:  General Glass Blowing
Description:   Sturdy, molded, clear-glass block
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Cylindrical glass tanks for the development of 1x3" to 10x20cm plates
Supplier:  General Glass Blowing
Description:   These TLC developing chambers are designed for 10cm plates.
Supplier:  General Glass Blowing
Description:   The Latch-Lid ChromatoTank is a versatile laboratory product
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 12
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping