Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
200-4298 200-4298
CA200-4298EA 8179.44 CAD
10802-062 CA200-4298
Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR