Anti-GST Goat Polyclonal Antibody (Agarose)

Supplier: Rockland Immunochemicals
200-119-200
CA200-119-200EA 8179.44 CAD
CA200-119-200
Anti-GST Goat Polyclonal Antibody (Agarose)
Antibodies
Type: Primary
Antigen: GST (Glutathione-S-Transferase)
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Goat
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR