Anti-Biotin Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
200-1298 200-1298
CA200-1298EA 8179.44 CAD
CA200-1298 10802-060
Anti-Biotin Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: Biotin
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR