Anti-IgG Donkey Polyclonal Antibody

Supplier: Rockland Immunochemical
211-7103
CA211-7103EA 1210.9 CAD
CA211-7103
Anti-IgG Donkey Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR