Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
310-1102 310-1102
CA310-1102EA 581.41 CAD
CA310-1102 10802-684
Anti-IgG Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR