Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
303-4104 303-4104
CA303-4104EA 584.82 CAD
10802-670 CA303-4104
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR