Anti-Luciferase Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-150
CA200-106-150EA 5384.89 CAD
CA200-106-150
Anti-Luciferase Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Firefly
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR