Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-4397 200-4397
CA200-4397EA 8179.44 CAD
10801-676 CA200-4397
Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Tag
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR