National LoVial™ Crimp Top Vials with Wide Opening, Thermo Scientific
Supplier: Thermo Scientific
C4011-2
66030-122CS 360 CAD
66030-122
National LoVial™ Crimp Top Vials with Wide Opening, Thermo Scientific
Vials
Thermo Scientific National Target LoVial Crimp Top Vials are designed with a wide opening for easier filling.
Vials fit most autosamplers that accommodate the 12×32 mm vial format. Made of borosilicate clear glass Type 1, Class A or 51A amber glass. Uniformly flat bottom for security with inserts. I-D vials feature write-on patch with graduations. Target Colorband crimp top vials feature color-coded graduations and patch. Vials with an 11 mm crimp finish require aluminum seal closures. Crimp/snap vials can be used with either 11 mm aluminum seals or Snap-It Snap Caps.
Keep all vials and compatible crimp seals handy and dust-free with Thermo Scientific National Unassembled Target LoVial Wide-Opening Vial Kits, which include matched quantities of vials and silver aluminum seals with prefitted septa. Two-compartment benchtop storage tray.