Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
210-401-319
CA10169-880EA 757.91 CAD
CA10169-880
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR