Anti-IgG Goat Polyclonal Antibody (Cy5.5®)

Supplier: Rockland Immunochemicals
606-113-129
CA606-113-129EA 630.83 CAD
CA606-113-129
Anti-IgG Goat Polyclonal Antibody (Cy5.5®)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR