Anti-Plasminogen Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
200101208-0100 200-101-208-0100
CARL200101208EA 947.07 CAD
RL200101208010 CARL200101208
Anti-Plasminogen Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Plasminogen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR