Anti-NAM-DH Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-101-228-0100 200101228-0100
CARL200101228EA 947.3 CAD
RL200101228010 CARL200101228
Anti-NAM-DH Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NAM-DH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Pseudomonas
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR