Anti-ACH Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
200101232-0100 200-101-232-0100
CARL200101232EA 947.3 CAD
CARL200101232 RL200101232010
Anti-ACH Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR