Anti-CPY Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-135-0100 200401135-0100
CARL200401135EA 947.3 CAD
CARL200401135 RL200401135010
Anti-CPY Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR