Anti-ADA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200401140-0100
CARL200401140EA 947.3 CAD
CARL200401140
Anti-ADA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR