Anti-D TMP Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
200601199-0100
CARL200601199EA 947.3 CAD
CARL200601199
Anti-D TMP Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: D TMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR