Anti-FGA Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200106240-0100
CA11028-870EA 1125.12 CAD
CA11028-870
Anti-FGA Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR