Anti-TRP1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200406139-0100
CA11028-886EA 1125.43 CAD
CA11028-886
Anti-TRP1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: TRP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR