Anti-ALPI Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200406142-0100
CA11028-892EA 1125.43 CAD
CA11028-892
Anti-ALPI Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR