Anti-ALDH1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4644-0100 200-4644-0100
CA11029-292EA 949.55 CAD
RL20046440100 CA11029-292
Anti-ALDH1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ALDH1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR