Anti-HK Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-4659-0100
CA11029-308EA 949.55 CAD
CA11029-308
Anti-HK Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: HK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR