Anti-PDH Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4660-0100 200-4660-0100
CA11029-310EA 949.55 CAD
CA11029-310 RL20046600100
Anti-PDH Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PDH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR