Anti-PDH Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4660S 200-4660S
CA11029-078EA 258.98 CAD
CA11029-078 RL2004660S
Anti-PDH Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PDH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR