Anti-CES Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4669-0100 200-4669-0100
CA11029-320EA 949.55 CAD
CA11029-320 RL20046690100
Anti-CES Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CES
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR